Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119564.2 | XP_013975039.1 | 3 - 4 | 724 | 106 | |
XM_014119565.2 | XP_013975040.1 | 3 - 4 | 259 | 106 | ||
XM_022427982.1 | XP_022283690.1 | 3 - 4 | 635 | 106 | ||
XM_845204.5 | XP_850297.1 | 3 - 4 | 634 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |