Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626854.3 | XP_005626911.2 | 3 - 4 | 286 | 178 | |
XM_014117879.2 | XP_013973354.1 | 3 - 4 | 355 | 178 | ||
XM_022425638.1 | XP_022281346.1 | 4 - 5 | 460 | 178 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |