Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628352.3 | XP_005628409.1 | 2 | 944 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |