Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003435139.4 | XP_003435187.1 | 3 - 4 | 1280 | 169 | |
XM_014115917.2 | XP_013971392.1 | 4 - 5 | 709 | 169 | ||
XM_022422037.1 | XP_022277745.1 | 2 - 3 | 605 | 169 | ||
XM_547830.6 | XP_547830.2 | 4 - 5 | 681 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |