Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620387.2 | XP_005620444.1 | 432 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |