Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003432576.5 | XP_003432624.2 | 3 - 4 | 461 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |