Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630498.3 | XP_005630555.1 | 2 | 1877 | 79 | |
XM_014120229.2 | XP_013975704.1 | 2 | 1691 | 79 | ||
XM_540204.6 | XP_540204.1 | 2 | 1873 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |