Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FILIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639388.3 | XP_005639445.1 | 3780 | 94 | ||
XM_022413927.1 | XP_022269635.1 | 3432 | 94 | |||
XM_022413928.1 | XP_022269636.1 | 3209 | 94 | |||
XM_022413929.1 | XP_022269637.1 | 3204 | 94 | |||
XM_022413930.1 | XP_022269638.1 | 3084 | 94 | |||
XM_022413931.1 | XP_022269639.1 | 3118 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |