Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR51V20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639792.1 | XP_003639840.1 | 1 | 790 | 154 |
Gene Symbol: cOR51V3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407452.1 | XP_022263160.1 | 1 | 628 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |