Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640218.4 | XP_003640266.1 | 1 - 2 | 910 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |