Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433161.4 | XP_003433209.1 | 3 | 810 | 69 | |
XM_005634552.3 | XP_005634609.1 | 4 | 901 | 69 | ||
XM_022408730.1 | XP_022264438.1 | 2 | 726 | 69 | ||
GenBank mRNA | U67200.1 | - | 1 | 176 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Both primers and probe map within a single exon |