Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_857165.5 | XP_862258.2 | 592 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Inventoried |