Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195844.1 | NP_001182773.1 | 1 | 848 | 67 | |
XM_005618866.3 | XP_005618923.1 | 2 | 1461 | 67 | ||
XM_005618867.3 | XP_005618924.1 | 2 | 1581 | 67 | ||
XM_005618868.3 | XP_005618925.1 | 2 | 1461 | 67 | ||
XM_022417328.1 | XP_022273036.1 | 2 | 1461 | 67 | ||
XM_022417329.1 | XP_022273037.1 | 2 | 1461 | 67 | ||
XM_022417330.1 | XP_022273038.1 | 2 | 1461 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |