Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197029.1 | NP_001183958.1 | 4 - 5 | 823 | 74 | |
GenBank mRNA | FJ906970.1 | - | 4 - 5 | 823 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |