Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197167.1 | NP_001184096.1 | 3 - 4 | 599 | 80 | |
XM_005615763.2 | XP_005615820.1 | 4 - 5 | 770 | 80 | ||
XM_005615764.2 | XP_005615821.1 | 4 - 5 | 765 | 80 | ||
GenBank mRNA | HM135453.1 | - | 3 - 4 | 599 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |