Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205203.1 | NP_001192132.1 | 3 - 4 | 394 | 86 | |
XM_022406260.1 | XP_022261968.1 | 4 - 5 | 1300 | 86 | ||
XM_022406261.1 | XP_022261969.1 | 4 - 5 | 563 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |