Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252257.1 | NP_001239186.1 | 1 | 221 | 108 | |
NM_001252258.1 | NP_001239187.1 | 1 | 130 | 108 | ||
XM_022423989.1 | XP_022279697.1 | 1 | 136 | 108 | ||
XM_022423990.1 | XP_022279698.1 | 1 | 87 | 108 | ||
XM_022423991.1 | XP_022279699.1 | 267 | 108 | |||
XM_022423992.1 | XP_022279700.1 | 1 | 201 | 108 | ||
GenBank mRNA | DT536457.1 | - | 1 | 137 | 108 | |
GR888023.1 | - | 1 | 198 | 108 | ||
GR892750.1 | - | 221 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |