Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270887.1 | NP_001257816.1 | 9 - 10 | 980 | 76 | |
XM_022423975.1 | XP_022279683.1 | 12 - 13 | 1217 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |