Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111456.2 | XP_013966931.1 | 2 - 3 | 634 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Inventoried Probe spans exons |