Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244079.1 | NP_001231008.1 | 1 | 547 | 61 | |
GenBank mRNA | AF527628.1 | - | 1 | 547 | 61 | |
JI879777.1 | - | 1 | 231 | 61 | ||
JP062602.1 | - | 1 | 231 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |