Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246695.1 | NP_001233624.1 | 4 - 5 | 873 | 63 | |
XM_007649755.1 | XP_007647945.1 | 7 - 8 | 1390 | 63 | ||
XM_007649756.1 | XP_007647946.1 | 6 - 7 | 1181 | 63 | ||
XM_007649757.1 | XP_007647947.1 | 6 - 7 | 1112 | 63 | ||
XM_007649758.1 | XP_007647948.1 | 6 - 7 | 1295 | 63 | ||
XM_007649759.1 | XP_007647949.1 | 7 - 8 | 1231 | 63 | ||
GenBank mRNA | AY117538.1 | - | 4 - 5 | 873 | 63 | |
JI890368.1 | - | 4 - 5 | 944 | 63 | ||
JP037518.1 | - | 4 - 5 | 944 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |