Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246714.1 | NP_001233643.1 | 4 - 5 | 365 | 72 | |
GenBank mRNA | AB271736.1 | - | 4 - 5 | 365 | 72 | |
JI875398.1 | - | 4 - 5 | 451 | 72 | ||
JP037636.1 | - | 4 - 5 | 451 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |