Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511282.2 | XP_003511330.1 | 2 - 3 | 311 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Probe spans exons |