Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650149.1 | XP_007648339.1 | 4 - 5 | 692 | 90 | |
XM_007650150.1 | XP_007648340.1 | 4 - 5 | 1550 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Protease Probe spans exons |