Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cant1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498816.2 | XP_003498864.1 | 3 - 4 | 1154 | 79 | |
GenBank mRNA | JI874637.1 | - | 1 - 2 | 197 | 79 | |
JP055533.1 | - | 1 - 2 | 197 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |