Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502321.2 | XP_003502369.1 | 2 - 3 | 242 | 63 | |
GenBank mRNA | JI869557.1 | - | 2 - 3 | 229 | 63 | |
JP048522.1 | - | 2 - 3 | 229 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |