Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504693.2 | XP_003504741.1 | 2 - 3 | 471 | 97 | |
GenBank mRNA | JP061365.1 | - | 2 - 3 | 481 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |