Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508170.2 | XP_003508218.1 | 2 - 3 | 724 | 70 | |
XM_007648792.1 | XP_007646982.1 | 2 - 3 | 728 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |