Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643795.1 | XP_007641985.1 | 3 - 4 | 420 | 104 | |
GenBank mRNA | JI863231.1 | - | 1 - 2 | 117 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |