Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501218.2 | XP_003501266.1 | 8 - 9 | 1107 | 72 | |
XM_007641906.1 | XP_007640096.1 | 8 - 9 | 943 | 72 | ||
XM_007641907.1 | XP_007640097.1 | 8 - 9 | 1107 | 72 | ||
GenBank mRNA | JP061130.1 | - | 3 - 4 | 378 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |