Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505102.2 | XP_003505150.1 | 1 - 2 | 214 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |