Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648738.1 | XP_007646928.1 | 3 - 4 | 516 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Protease Detect Genomic DNA Probe spans exons |