Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514828.2 | XP_003514876.1 | 2 - 3 | 310 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |