Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647389.1 | XP_007645579.1 | 2 - 3 | 281 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Probe spans exons |