Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A3galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500624.1 | XP_003500672.1 | 3 - 4 | 193 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |