Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503244.2 | XP_003503292.1 | 5 - 6 | 1549 | 112 | |
XM_007643848.1 | XP_007642038.1 | 5 - 6 | 1554 | 112 | ||
GenBank mRNA | JP044056.1 | - | 4 - 5 | 541 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |