Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647874.1 | XP_007646064.1 | 1 - 2 | 105 | 94 | |
GenBank mRNA | JP062965.1 | - | 1 - 2 | 105 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |