Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513016.2 | XP_003513064.1 | 2 - 3 | 844 | 75 | |
GenBank mRNA | JI881971.1 | - | 2 - 3 | 866 | 75 | |
JP039088.1 | - | 2 - 3 | 866 | 75 |