Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644252.1 | XP_007642442.1 | 4 - 5 | 7026 | 74 | |
XM_007644253.1 | XP_007642443.1 | 5 - 6 | 6835 | 74 | ||
GenBank mRNA | JI875085.1 | - | 1 - 2 | 2084 | 74 | |
JP051018.1 | - | 1 - 2 | 2084 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |