Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633880.1 | XP_007632070.1 | 7 - 8 | 2030 | 55 | |
XM_007633884.1 | XP_007632074.1 | 7 - 8 | 1720 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |