Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rimkla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642478.1 | XP_007640668.1 | 4 - 5 | 485 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |