Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dolk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513968.2 | XP_003514016.1 | 1 | 549 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Both primers and probe map within a single exon |