Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505511.2 | XP_003505559.2 | 2 - 3 | 267 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |