Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502890.2 | XP_003502938.1 | 4 - 5 | 351 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |