Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512854.2 | XP_003512902.1 | 5 - 6 | 626 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |