Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495051.1 | XP_003495099.1 | 6 - 7 | 945 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |