Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507493.2 | XP_003507541.1 | 3 - 4 | 1252 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |