Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509272.2 | XP_003509320.1 | 1 - 2 | 155 | 106 | |
GenBank mRNA | JI891021.1 | - | 1 - 2 | 126 | 106 | |
JP038795.1 | - | 1 - 2 | 126 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |