Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495127.2 | XP_003495175.1 | 2 - 3 | 272 | 114 | |
GenBank mRNA | JI877023.1 | - | 2 - 3 | 238 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |