Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496566.2 | XP_003496614.1 | 3 - 4 | 461 | 66 | |
GenBank mRNA | JI868866.1 | - | 2 - 3 | 346 | 66 | |
JP050971.1 | - | 2 - 3 | 346 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |